Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Wheaton
®
Leak Resistant Bottle
Products
Product Category
bottles (51)
Capacity (mL)
Diameter (mm)
Bottle material
Color
Vial Bottle Material
Vial Bottle Color
Filter Products
Showing 1-20 of 51
Page 1 of 3
Page 1 of 3
Compare
Product Number
Description
Pricing
DWK209550
Wheaton
®
Leak Resistant Bottle,
capacity 1000 mL, high-density polyethylene bottle, natural bottle, wide-mouth bottle, bottle diam. × H 91 mm × 199 mm, 63-415
Expand
View Pricing
DWK209041
Wheaton
®
Leak Resistant Bottle,
capacity 4 mL, high-density polyethylene bottle, narrow-mouth bottle, natural bottle, bottle diam. × H 16 mm × 39 mm, 13-425
Expand
View Pricing
DWK209164
Wheaton
®
Leak Resistant Bottle,
capacity 15 mL, polypropylene bottle, natural bottle, narrow-mouth bottle, bottle diam. × H 25 mm × 56 mm, 20-410
Expand
View Pricing
DWK209548
Wheaton
®
Leak Resistant Bottle,
capacity 250 mL, high-density polyethylene bottle, natural bottle, wide-mouth bottle, bottle diam. × H 61 mm × 125 mm, 43-140
Expand
View Pricing
DWK209121
Wheaton
®
Leak Resistant Bottle,
capacity 4 mL, high-density polyethylene bottle, amber bottle, narrow-mouth bottle, bottle diam. × H 16 mm × 38 mm, 13-425
Expand
View Pricing
DWK209665
Wheaton
®
Leak Resistant Bottle,
capacity 30 mL, polypropylene bottle, natural bottle, wide-mouth bottle, bottle diam. × H 33 mm × 69 mm, 28-410
Expand
View Pricing
DWK209044
Wheaton
®
Leak Resistant Bottle,
capacity 15 mL, high-density polyethylene bottle, natural bottle, narrow-mouth bottle, bottle diam. × H 25 mm × 56 mm, 20-410
Expand
View Pricing
DWK209124
Wheaton
®
Leak Resistant Bottle,
capacity 15 mL, high-density polyethylene bottle, amber bottle, narrow-mouth bottle, bottle diam. × H 25 mm × 56 mm, 20-410
Expand
View Pricing
DWK209626
Wheaton
®
Leak Resistant Bottle,
capacity 60 mL, high-density polyethylene bottle, amber bottle, wide-mouth bottle, bottle diam. × H 39 mm × 88 mm, 28-410
Expand
View Pricing
DWK209043
Wheaton
®
Leak Resistant Bottle,
capacity 8 mL, high-density polyethylene bottle, natural bottle, narrow-mouth bottle, bottle diam. × H 25 mm × 43 mm, 20-410
Expand
View Pricing
DWK209050
Wheaton
®
Leak Resistant Bottle,
capacity 1000 mL, high-density polyethylene bottle, natural bottle, narrow-mouth bottle, bottle diam. × H 91 mm × 210 mm, 38-430
Expand
View Pricing
DWK209667
Wheaton
®
Leak Resistant Bottle,
capacity 125 mL, polypropylene bottle, natural bottle, wide-mouth bottle, bottle diam. × H 51 mm × 98 mm, 38-410
Expand
View Pricing
DWK209545
Wheaton
®
Leak Resistant Bottle,
capacity 30 mL, high-density polyethylene bottle, natural bottle, wide-mouth bottle, bottle diam. × H 33 mm × 69 mm, 28-410
Expand
View Pricing
DWK209627
Wheaton
®
Leak Resistant Bottle,
capacity 125 mL, high-density polyethylene bottle, amber bottle, wide-mouth bottle, bottle diam. × H 51 mm × 98 mm, 38-410
Expand
View Pricing
DWK209045
Wheaton
®
Leak Resistant Bottle,
capacity 30 mL, high-density polyethylene bottle, natural bottle, narrow-mouth bottle, bottle diam. × H 32 mm × 69 mm, 20-410
Expand
View Pricing
DWK209625
Wheaton
®
Leak Resistant Bottle,
capacity 30 mL, high-density polyethylene bottle, amber bottle, wide-mouth bottle, bottle diam. × H 33 mm × 69 mm, 28-410
Expand
View Pricing
DWK209170
Wheaton
®
Leak Resistant Bottle,
capacity 1000 mL, polypropylene bottle, natural bottle, narrow-mouth bottle, bottle diam. × H 91 mm × 210 mm, 38-430
Expand
View Pricing
DWK209048
Wheaton
®
Leak Resistant Bottle,
capacity 250 mL, high-density polyethylene bottle, natural bottle, narrow-mouth bottle, bottle diam. × H 61 mm × 129 mm, 24-410
Expand
View Pricing
DWK209161
Wheaton
®
Leak Resistant Bottle,
capacity 4 mL, polypropylene bottle, natural bottle, narrow-mouth bottle, bottle diam. × H 16 mm × 38 mm, 13-425
Expand
View Pricing
DWK208925
Wheaton
®
Leak Resistant Bottle,
capacity 30 mL, low-density polyethylene bottle, natural bottle, narrow-mouth bottle, bottle diam. × H 32 mm × 69 mm, 20-410
Expand
View Pricing
Page 1 of 3
Page 1 of 3