Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
Vacuum filtration apparatus accessories
Synonym(s):
Whatman
®
Vacuum filtration apparatus accessories
Products
Product Category
holders (7)
tweezers (1)
Manufacturer
Housing Material
Package Size
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
WHA10471700
Whatman
®
Vacuum filtration apparatus accessories,
SV 006 vacuum tubing, 1 meter length
Expand
View Pricing
WHA10477600
Whatman
®
Vacuum filtration apparatus accessories,
SF 100 suction flask, 1000 mL with tubing nozzle
Expand
View Pricing
WHA10477602
Whatman
®
Vacuum filtration apparatus accessories,
PZ 001 tweezers, stainless steel
Expand
View Pricing
WHA10477601
Whatman
®
Vacuum filtration apparatus accessories,
WT 100 Witt′s flask, with tubing nozzle, capacity
Expand
View Pricing
WHA10464103
Whatman
®
Vacuum filtration apparatus accessories,
ML 050/0/03 steel frit with ring, pack of 1
Expand
View Pricing
WHA10440001
Whatman
®
Vacuum filtration apparatus accessories,
MV050/0/01 stainless steel funnel complete for MV050 series, capacity 500 mL, pack of 1
Expand
View Pricing
WHA10440003
Whatman
®
Vacuum filtration apparatus accessories,
MV050/0/03 sieve for MV050 series, pack of 1
Expand
View Pricing
WHA10440102
Whatman
®
Vacuum filtration apparatus accessories,
MV 050/1/05 PTFE gasket for MV050 series, W × L × D 50 mm × 43 mm × 0.5 mm, pack of 1
Expand
View Pricing
WHA10464100
Whatman
®
Vacuum filtration apparatus accessories,
ML 050/0 in-line filter holder, stainless steel, pack of 1
Expand
View Pricing