Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
UNIPLATE microplates
Products
Product Category
plates (6)
Manufacturer
Binding Type
Volume (ml)
Volume (µL)
Wells
Working Volume (ml)
Color
Package Size
Special Grade
Natural (3)
Sterility
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
WHA77015200
Whatman
®
UNIPLATE microplates,
96 well, round bottom, 2ml volume, PP, pkg of 25 ea
Expand
View Pricing
WHA77015102
Whatman
®
UNIPLATE microplates,
24 well, round well bottom, well volume 10 mL, natural polypropylene
Expand
View Pricing
WHA77011651
Whatman
®
UNIPLATE microplates,
96 well, flat bottom, well volume 650 μL, clear polystyrene
Expand
View Pricing
WHA77013250
Whatman
®
UNIPLATE microplates,
96 well, "V" bottom, well volume 250 μL, white polystyrene
Expand
View Pricing
WHA77015110
Whatman
®
UNIPLATE microplates,
24 well, round well bottom, irradiated with lid, well volume 10 mL, natural polypropylene
Expand
View Pricing
WHA77015500
Whatman
®
UNIPLATE microplates,
48 well, flat bottom, well volume 5 mL, natural polypropylene
Expand
View Pricing