Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
mixed cellulose ester ME 27 filter discs 0.8 μm pore size
Products
Product Category
filter discs (7)
Manufacturer
Diameter (mm)
Color
Package Size
Pore size (µm)
Sterility
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
WHA10400914
Whatman
®
mixed cellulose ester ME 27 filter discs 0.8 μm pore size,
white (no grid), hydrophilic, nonsterile, interleaving paper, 100 EA, 100 ea, 50 mm diam
Expand
View Pricing
WHA10400912
Whatman
®
mixed cellulose ester ME 27 filter discs 0.8 μm pore size,
white (no grid), hydrophilic, nonsterile, interleaving paper, 100 EA, 100 ea, 47 mm diam
Expand
View Pricing
WHA10400906
Whatman
®
mixed cellulose ester ME 27 filter discs 0.8 μm pore size,
white (no grid), hydrophilic, nonsterile, interleaving paper, 100 EA, 100 ea, 25 mm diam
Expand
View Pricing
WHA10408970
Whatman
®
mixed cellulose ester ME 27 filter discs 0.8 μm pore size,
white, hydrophilic, black grid (3.1 mm), sterile, interleaving paper, 100 EA, 100 ea, 47 mm diam
Expand
View Pricing
WHA10400909
Whatman
®
mixed cellulose ester ME 27 filter discs 0.8 μm pore size,
white (no grid), hydrophilic, nonsterile, interleaving paper, 100 EA, 100 ea, 37 mm diam
Expand
View Pricing
WHA10409970
Whatman
®
mixed cellulose ester ME 27 filter discs 0.8 μm pore size,
white, hydrophilic, black grid (3.1 mm), sterile, pad, 100 EA, 100 ea, 47 mm diam
Expand
View Pricing
WHA10400921
Whatman
®
mixed cellulose ester ME 27 filter discs 0.8 μm pore size,
white (no grid), hydrophilic, nonsterile, interleaving paper, 50 EA, 50 ea, 100 mm diam
Expand
View Pricing