Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
Mini-UniPrep
®
syringeless filters with slit septa
Products
Product Category
filters (22)
syringe filters (9)
Manufacturer
Cap
Housing Material
Membrane Material
Pore size (µm)
Sterility
Filter Products
Showing 1-20 of 22
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
WHAUS503NPUORG
Whatman
®
Mini-UniPrep
®
syringeless filters with slit septa,
PTFE, 0.45 μm, 100/pk or 1000/pk
Expand
View Pricing
WHAUS503NPEAQU
Whatman
®
Mini-UniPrep
®
syringeless filters with slit septa,
PVDF, 0.2 μm, 1000/pk
Expand
View Pricing
WHAUS503NPEORG
Whatman
®
Mini-UniPrep
®
syringeless filters with slit septa,
PTFE, 0.2 μm, 1000/pk
Expand
View Pricing
WHAUS503NPUAQU
Whatman
®
Mini-UniPrep
®
syringeless filters with slit septa,
PVDF, 0.45 μm, 1000/pk
Expand
View Pricing
WHAUS203NPUORG
Whatman
®
Mini-UniPrep
®
syringeless filters with slit septa,
PTFE, 0.45 μm, 100/pk
Expand
View Pricing
WHAUS503NPEPP
Whatman
®
Mini-UniPrep
®
syringeless filters with slit septa,
PP, 0.2 μm, 1000/pk
Expand
View Pricing
WHAUS203NPEORG
Whatman
®
Mini-UniPrep
®
syringeless filters with slit septa,
PTFE, 0.2 μm, 100/pk
Expand
View Pricing
WHAUS203NPEPP
Whatman
®
Mini-UniPrep
®
syringeless filters with slit septa,
PP, 0.2 μm, 100/pk
Expand
View Pricing
WHAUS203NPUAQU
Whatman
®
Mini-UniPrep
®
syringeless filters with slit septa,
PVDF, 0.45 μm, 100/pk
Expand
View Pricing
WHAUS203NPEAQU
Whatman
®
Mini-UniPrep
®
syringeless filters with slit septa,
PVDF, 0.2 μm, 100/pk
Expand
View Pricing
WHAUS203NPENYL
Whatman
®
Mini-UniPrep
®
syringeless filters with slit septa,
Nylon, 0.2 μm, 100/pk
Expand
View Pricing
WHAUS203NPUNYL
Whatman
®
Mini-UniPrep
®
syringeless filters with slit septa,
Nylon, 0.45 μm, 100/pk
Expand
View Pricing
WHAUS203NPUDPP
Whatman
®
Mini-UniPrep
®
syringeless filters with slit septa,
PP, 0.45 μm, 100/pk
Expand
View Pricing
WHAUS203NPUPP
Whatman
®
Mini-UniPrep
®
syringeless filters with slit septa,
PP, 0.45 μm, 100/pk
Expand
View Pricing
WHAUS203NPEPES
Whatman
®
Mini-UniPrep
®
syringeless filters with slit septa,
PES, 0.2 μm, 100/pk
Expand
View Pricing
WHAUS203NPUGMF
Whatman
®
Mini-UniPrep
®
syringeless filters with slit septa,
GMF, 0.45 μm, 100/pk
Expand
View Pricing
WHAUS503NPENYL
Whatman
®
Mini-UniPrep
®
syringeless filters with slit septa,
nylon, 0.2 μm, 1000/pk
Expand
View Pricing
WHAUS503NPUGMF
Whatman
®
Mini-UniPrep
®
syringeless filters with slit septa,
GMF, 0.45 μm, 1000/pk
Expand
View Pricing
WHAUS203NPUPES
Whatman
®
Mini-UniPrep
®
syringeless filters with slit septa,
PES, 0.45 μm, 100/pk
Expand
View Pricing
WHAUS503NPUPP
Whatman
®
Mini-UniPrep
®
syringeless filters with slit septa,
PP, 0.45 μm, 1000/pk
Expand
View Pricing
Page 1 of 2
Page 1 of 2