Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
glass microfiber membrane holder
Products
Product Category
holders (8)
Manufacturer
Capacity (mL)
Diameter (mm)
Volume (ml)
Package Size
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
WHA1980001
Whatman
®
glass microfiber membrane holder,
Stainless Steel Syringe Filter Type, diam. 13 mm
Expand
View Pricing
WHA1961054
Whatman
®
glass microfiber membrane holder,
Glass Reservoir, volume 300 mL, diam. 47 mm
Expand
View Pricing
WHA1960004
Whatman
®
glass microfiber membrane holder,
Glass Support, volume 300 mL, diam. 47 mm
Expand
View Pricing
WHA1960009
Whatman
®
glass microfiber membrane holder,
Glass Support, volume 1000 mL, diam. 90 mm
Expand
View Pricing
WHA1960054
Whatman
®
glass microfiber membrane holder,
Stainless Steel Support, volume 300 mL, diam. 47 mm
Expand
View Pricing
WHA1960002
Whatman
®
glass microfiber membrane holder,
Glass Reservoir, volume 25 mL, diam. 25 mm
Expand
View Pricing
WHA1980002
Whatman
®
glass microfiber membrane holder,
Stainless Steel Syringe Filter Type, diam. 25 mm
Expand
View Pricing
WHA1960052
Whatman
®
glass microfiber membrane holder,
stainless steel membrane holder, capacity 25 mL
Expand
View Pricing
WHA1960032
Whatman
®
glass microfiber membrane holder,
Glass Support, volume 50 mL, diam. 25 mm
Expand
View Pricing