Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Western Blot Box
Products
Product Category
boxes (9)
Color
Manufacturer
Package Size
Size
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
Z742099
Western Blot Box,
black, size 8.9 cm × 6.5 cm × 2.5 cm, pk of 5
Expand
View Pricing
Z742098
Western Blot Box,
clear, size 8.9 cm × 6.5 cm × 2.5 cm, pk of 5
Expand
View Pricing
Z742100
Western Blot Box,
clear, size 9.5 cm × 3.0 cm × 1.6 cm, pk of 5
Expand
View Pricing
Z742096
Western Blot Box,
clear, size 7.3 cm × 3.0 cm × 1.9 cm, pk of 5
Expand
View Pricing
Z742091
Western Blot Box,
size large, clear, size 11.7 cm × 8.9 cm × 2.8 cm, pk of 5
Expand
View Pricing
Z742097
Western Blot Box,
clear, size 7.3 cm × 5.1 cm × 3.2 cm, pk of 5
Expand
View Pricing
Z742095
Western Blot Box,
size XXL, size 15.2 cm × 10.2 cm × 5.1 cm, pk of 5
Expand
View Pricing
Z742092
Western Blot Box,
size (square), size 11.4 cm × 11.4 cm × 3.2 cm, pk of 5
Expand
View Pricing
Z742093
Western Blot Box,
size (square), size 11.4 cm × 11.4 cm × 3.2 cm, pk of 5
Expand
View Pricing