Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Wash-N-Dry
™
coverslip rack
Products
Product Category
racks (5)
Color
Manufacturer
Package Size
Special Grade
Natural (1)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
Z688568
Wash-N-Dry
™
coverslip rack,
holds 10 slipcover sizes including 18, 22 and 25 mm, natural
Expand
View Pricing
Z743685
Wash-N-Dry
™
coverslip rack,
XL, blue, holds 30 slipcover sizes including 18, 22 and 25 mm
Expand
View Pricing
Z743684
Wash-N-Dry
™
coverslip rack,
blue, holds 10 slipcover sizes including 18, 22 and 25 mm
Expand
View Pricing
Z743686
Wash-N-Dry
™
coverslip rack,
XL, yellow, holds 30 slipcover sizes including 18, 22 and 25 mm
Expand
View Pricing
Z743687
Wash-N-Dry
™
coverslip rack,
yellow, holds all slipcover sizes including 18, 22 and 25 mm
Expand
View Pricing