
Warfarin

Synonym(s):
4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin, Coumafene
Empirical Formula (Hill Notation):
C19H16O4
CAS Number:
Molecular Weight:
308.33
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing