Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Urease from
Canavalia ensiformis
(Jack bean)
CAS Number:
9002-13-5
EC Number:
232-656-0
Products
Product Category
enzymes (2)
Brand
Sigma-Aldrich (7)
Biological Source
Color
Physical Form
Quality Segment
Application
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
U1500
Urease from
Canavalia ensiformis
(Jack bean),
Type III, powder, 15,000-50,000 units/g solid
Expand
View Pricing
U4002
Urease from
Canavalia ensiformis
(Jack bean),
Type IX, powder, 50,000-100,000 units/g solid
Expand
View Pricing
94280
Urease from
Canavalia ensiformis
(Jack bean),
powder, ~1 U/mg
Expand
View Pricing
U1875
Urease from
Canavalia ensiformis
(Jack bean),
Type III, glycerol solution, 500-800 units/mL
Expand
View Pricing
U0251
Urease from
Canavalia ensiformis
(Jack bean),
Type C-3, powder, ≥600,000 units/g solid
Expand
View Pricing
94281
Urease from
Canavalia ensiformis
(Jack bean),
powder, white, ~8 U/mg
Expand
View Pricing
U7752
Urease from
Canavalia ensiformis
(Jack bean),
Expand
View Pricing