Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Upconversion Nanoparticles
Products
Brand
Sigma-Aldrich (16)
Quality Segment
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
926620
Upconversion Nanoparticles,
NaYF
4
:Yb,Er@NaYF
4
, Oil soluble core-shell, fluorescence λ
ex
980, green light
Expand
View Pricing
926590
Upconversion Nanoparticles,
NaYF
4
-Yb,Tm@NaYF
4
, PEG-COOH modified core-shell, fluorescence λ
ex
980, blue light
Expand
View Pricing
926558
Upconversion Nanoparticles,
NaYF4-Yb,Tm@NaYF4, PEG-NH2 modified core-shell ucnps,980 excitation, blue light
Expand
View Pricing
926604
Upconversion Nanoparticles,
NaYF
4
-Yb,Er@NaYF
4
, PEG-COOH modified core-shell, fluorescence λ
ex
980 nm, green light
Expand
View Pricing
926612
Upconversion Nanoparticles,
NaYF
4
,Yb,Tm@NaYF
4
,Yb,Nd, Oil soluble core shell upconversion nanoparticle, 808 excited, blue light
Expand
View Pricing
926515
Upconversion Nanoparticles,
Silica coated NaYF
4
-Yb,Er@NaYF
4
, fluorescence λ
ex
980 nm, green light
Expand
View Pricing
926671
Upconversion Nanoparticles,
Silica coated NaYF
4
,Yb,Tm@NaYF4,Yb,Nd, 808 excited, blue light
Expand
View Pricing
926639
Upconversion Nanoparticles,
NaYF4:Yb,Tm@NaYF4, Oil soluble core shell, 980 excitation, blue light
Expand
View Pricing
926663
Upconversion Nanoparticles,
Silica coated NaYF4,Yb,Er@NaYF4,Yb,Nd, 808 excited, green
Expand
View Pricing
926574
Upconversion Nanoparticles,
NaYF
4
,Yb,Tm@NaYF
4
,Yb,Nd, PEG-COOH modified core-shell, fluorescence λ
ex
808 nm, blue light
Expand
View Pricing
926655
Upconversion Nanoparticles,
Silica coated NaYF
4
-Yb,Tm@NaYF4, 980 excitation, blue light
Expand
View Pricing
926647
Upconversion Nanoparticles,
NaYF
4
,Yb,Er@NaYF
4
,Yb,Nd, Oil soluble core shell,808 excitation, green
Expand
View Pricing
926523
Upconversion Nanoparticles,
NaYF
4
,Yb,Tm@NaYF
4
Yb, Nd, PEG-NH
2
modified core-shell, fluorescence λ
ex
808 nm, blue light
Expand
View Pricing
926531
Upconversion Nanoparticles,
NaYF
4
,Yb,Er@NaYF
4
,Yb,Nd, PEG-NH
2
modified core-shell, 808 excitation, green light
Expand
View Pricing
926566
Upconversion Nanoparticles,
NaYF
4
-Yb,Er@NaYF
4
, PEG-NH
2
modified core-shell, 980 excitation, green light
Expand
View Pricing
926582
Upconversion Nanoparticles,
NaYF
4
,Yb,Er@NaYF
4
,Yb,Nd, PEG-COOH modified core-shell, 808 excitation, green light
Expand
View Pricing