
U-73122 hydrate

Synonym(s):
1-[6-[((17β)-3-Methoxyestra-1,3,5[10]-trien-17-yl)amino]hexyl]-1H-pyrrole-2,5-dione
Empirical Formula (Hill Notation):
C29H40N2O3 · xH2O
CAS Number:
Molecular Weight:
464.64 (anhydrous basis)
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing