Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tungsten(VI) oxide
Synonym(s):
Tungstic anhydride
Linear Formula:
WO
3
CAS Number:
1314-35-8
Molecular Weight:
231.84
EC Number:
215-231-4
Products
Brand
Sigma-Aldrich (5)
Formula Weight
Physical Form
Purity
Quality Segment
Particle Size (µm)
Special Grade
Puriss (1)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
550086
Tungsten(VI) oxide,
nanopowder, <100 nm particle size (TEM)
Expand
View Pricing
204781
Tungsten(VI) oxide,
powder, 99.995% trace metals basis
Expand
View Pricing
232785
Tungsten(VI) oxide,
powder, ≤25 μm, ≥99% trace metals basis
Expand
View Pricing
95410
Tungsten(VI) oxide,
powder, puriss., 99.9%
Expand
View Pricing
774537
Tungsten(VI) oxide,
nanowires, diam. × L ~50 nm × 10 μm
Expand
View Pricing