Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tungsten carbide
Products
Manufacturer
Physical Form
Particle Size (µm)
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
GF08742539
Tungsten carbide,
powder, max. particle size 45 micron, weight 20 g, purity 99.5%
Expand
View Pricing
GF98618619
Tungsten carbide,
powder, 45 max. part. size (micron), weight 100 g, purity 99.5%
Expand
View Pricing
GF74580712
Tungsten carbide,
powder, 45 max. part. size (micron), weight 200 g, purity 99.5%
Expand
View Pricing
GF48047722
Tungsten carbide,
powder, max. particle size 45 micron, weight 50 g, purity 99.5%
Expand
View Pricing
GF28290438
Tungsten carbide,
powder, max. particle size 150 micron, weight 100 g, min. particle size 45 micron
Expand
View Pricing
GF36304400
Tungsten carbide,
powder, max. particle size 150 micron, weight 200 g, min. particle size 45 micron
Expand
View Pricing
GF40571412
Tungsten carbide,
powder, max. particle size 150 micron, weight 20 g, min. particle size 45 micron
Expand
View Pricing
GF71426325
Tungsten carbide,
powder, 150 max. part. size (micron), weight 50 g, min. particle size 45 micron
Expand
View Pricing