Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tubing quick-disconnects
Synonym(s):
Tubing quick-disconnects
Products
Brand
Sigma-Aldrich (1)
Manufacturer
Length (in)
Package Size
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
Z104485
Tubing quick-disconnects,
for tubing i.d., 1/8 ‑ 1/4 in.
Expand
View Pricing
Z104353
Tubing quick-disconnects,
for tubing i.d., 1/4 ‑ 5/16 in.
Expand
View Pricing
Z104361
Tubing quick-disconnects,
for tubing i.d., 3/8 ‑ 1/2in.
Expand
View Pricing
BAF197300001
Tubing quick-disconnects,
for ³/₁₆ to ¼ in. Tubing
Expand
View Pricing
BAF197300002
Tubing quick-disconnects,
for ¼ to ⅜ in. Tubing
Expand
View Pricing