Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Trypsin inhibitor from
Glycine max
(soybean)
Synonym(s):
SBTI
CAS Number:
9035-81-8
EC Number:
232-906-9
Products
Product Category
inhibitors (2)
proteases (1)
Brand
Sigma-Aldrich (3)
Biological Source
Physical Form
Product Line
Quality Segment
Sterilization Method
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
T6522
Trypsin inhibitor from
Glycine max
(soybean),
powder, BioReagent, suitable for cell culture
Expand
View Pricing
T6414
Trypsin inhibitor from
Glycine max
(soybean),
solution, sterile-filtered, suitable for, suitable for cell culture
Expand
View Pricing
93620
Trypsin inhibitor from
Glycine max
(soybean),
lyophilized powder, ~10000 U/mg
Expand
View Pricing