Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
TPP
®
tissue culture flasks
Products
Product Category
flasks (7)
Manufacturer
Binding Type
Capacity (mL)
Height (mm)
Length (mm)
Surface Area cm²
Working Volume (ml)
Cap
Package Size
Sterility
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
Z707562
TPP
®
tissue culture flasks,
capacity 410 mL, Tissue Culture (TC)-treated surface, with cap (filtered), case of 18 ea
Expand
View Pricing
Z707503
TPP
®
tissue culture flasks,
capacity 65 mL, Tissue Culture (TC)-treated surface, with cap, vented, case of 1000 ea
Expand
View Pricing
Z707546
TPP
®
tissue culture flasks,
capacity 65 mL, Tissue Culture (TC)-treated surface, with cap, filtered, case of 1000 ea
Expand
View Pricing
Z707554
TPP
®
tissue culture flasks,
capacity 165 mL, Tissue Culture (TC)-treated surface, with cap (filtered), case of 36 ea
Expand
View Pricing
Z707538
TPP
®
tissue culture flasks,
capacity 15 mL, Tissue Culture (TC)-treated surface, with cap (Filtered), case of 360 ea
Expand
View Pricing
Z707511
TPP
®
tissue culture flasks,
capacity 165 mL, Tissue Culture (TC)-treated surface, with cap, vented, case of 36 ea
Expand
View Pricing
Z707481
TPP
®
tissue culture flasks,
capacity 15 mL, Tissue Culture (TC)-treated surface, with cap, vented, case of 360 ea
Expand
View Pricing