Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
TPP
®
tissue culture dishes
Products
Product Category
dishes (4)
Manufacturer
Binding Type
Capacity (mL)
Diameter (mm)
Surface Area cm²
Package Size
Sterility
Sterilization Method
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
Z707686
TPP
®
tissue culture dishes,
dish capacity ~140 mL
Expand
View Pricing
Z707678
TPP
®
tissue culture dishes,
dish capacity ~40 mL
Expand
View Pricing
Z707651
TPP
®
tissue culture dishes,
dish capacity ~10 mL
Expand
View Pricing
Z707694
TPP
®
tissue culture dishes,
dish capacity ~350 mL
Expand
View Pricing