Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
TissueFab
®
bioink kit
Products
Brand
Sigma-Aldrich (6)
Color
Physical Form
Quality Segment
Application
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
926019
TissueFab
®
bioink kit,
(Gel)ma Fibronection -Vis/405 nm, low endotoxin
Expand
View Pricing
927074
TissueFab
®
bioink kit,
(Gel)ma Fibrin (Vis/405), low endotoxin
Expand
View Pricing
927066
TissueFab
®
bioink kit,
Fibronectin-UV/365nm
Expand
View Pricing
927058
TissueFab
®
bioink kit,
(Gel)ma Laminin -UV/365 nm, low endotoxin
Expand
View Pricing
926000
TissueFab
®
bioink kit,
(Gel)ma Laminin -Vis/405 nm, low endotoxin
Expand
View Pricing
927651
TissueFab
®
bioink kit,
(Gel)ma Fibrin (UV/365), low endotoxin
Expand
View Pricing