
Tigecycline

Synonym(s):
9-tert-Butylglycylamidominocycline, Tigecycline
Empirical Formula (Hill Notation):
C29H39N5O8
CAS Number:
Molecular Weight:
585.65
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing