Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tetrapropylammonium bromide
Linear Formula:
(CH
3
CH
2
CH
2
)
4
N(Br)
CAS Number:
1941-30-6
Molecular Weight:
266.26
EC Number:
217-727-6
Products
Product Category
salts (1)
Brand
Supelco (1)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
225568
Tetrapropylammonium bromide,
98%
Expand
View Pricing
88103
Tetrapropylammonium bromide,
suitable for ion pair chromatography, LiChropur
™
, ≥99.0% (AT)
Expand
View Pricing