Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tetrakis(dimethylamido)zirconium(IV)
Synonym(s):
Tetrakis(dimethylamino)zirconium(IV)
Linear Formula:
[(CH
3
)
2
N]
4
Zr
CAS Number:
19756-04-8
Molecular Weight:
267.53
Products
Brand
Sigma-Aldrich (1)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Core
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
579211
Tetrakis(dimethylamido)zirconium(IV),
electronic grade, ≥99.99% trace metals basis
Expand
View Pricing
669016
Tetrakis(dimethylamido)zirconium(IV),
packaged for use in deposition systems
Expand
View Pricing