Skip to Content
Merck
Tetraethylammonium acetate tetrahydrate 99%

Tetraethylammonium acetate tetrahydrate

Tetraethylammonium acetate tetrahydrate 99%
Linear Formula:
(C2H5)4N(OCOCH3) · 4H2O
CAS Number:
Molecular Weight:
261.36
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing