Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tetrabutylammonium chloride
Linear Formula:
[CH
3
(CH
2
)
3
]
4
NCl
CAS Number:
1112-67-0
Molecular Weight:
277.92
EC Number:
214-195-7
Products
Brand
Sigma-Aldrich (1)
Supelco (1)
Sustainability
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Purum (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
86870
Tetrabutylammonium chloride,
≥97.0% (NT)
Expand
View Pricing
86852
Tetrabutylammonium chloride,
suitable for ion pair chromatography, LiChropur
™
, ≥99.0%
Expand
View Pricing