Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tellurium
Empirical Formula (Hill Notation):
Te
CAS Number:
13494-80-9
Molecular Weight:
127.60
EC Number:
236-813-4
Products
Brand
Sigma-Aldrich (5)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
266418
Tellurium,
powder, −200 mesh, 99.8% trace metals basis
Expand
View Pricing
264865
Tellurium,
powder, −30 mesh, 99.997% trace metals basis
Expand
View Pricing
204544
Tellurium,
pieces, 99.999% trace metals basis
Expand
View Pricing
452378
Tellurium,
shot, ≤2 mm, 99.999% trace metals basis
Expand
View Pricing
263303
Tellurium,
granular, −5-+50 mesh, 99.99% trace metals basis
Expand
View Pricing