Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tedlar
®
SCV Gas Sampling Bag
Synonym(s):
Tedlar
®
Gas Sampling Bag
Products
Product Category
bags (7)
Brand
Supelco (7)
Agency
Length (in)
Quality Segment
Application
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
30272-U
Tedlar
®
SCV Gas Sampling Bag,
1L w/ Thermogreen
®
LB-2 Septa
Expand
View Pricing
30274-U
Tedlar
®
SCV Gas Sampling Bag,
5L w/ Thermogreen
®
LB-2 Septa
Expand
View Pricing
30284-U
Tedlar
®
SCV Gas Sampling Bag,
0.6L w/Thermogreen
®
LB-2 Septa
Expand
View Pricing
30273-U
Tedlar
®
SCV Gas Sampling Bag,
2L w/ Thermogreen
®
LB-2 Septa
Expand
View Pricing
30276-U
Tedlar
®
SCV Gas Sampling Bag,
10L w/ Thermogreen
®
LB-2 Septa
Expand
View Pricing
30244-U
Tedlar
®
SCV Gas Sampling Bag,
25L w/ Thermogreen
®
LB-2 Septa
Expand
View Pricing
30285-U
Tedlar
®
SCV Gas Sampling Bag,
3L w/ Thermogreen
®
LB-2 Septa
Expand
View Pricing