Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Synthware
™
solvent transfer/storage flask
Products
Product Category
flasks (6)
Manufacturer
Capacity (mL)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
SYNF484500
Synthware
™
solvent transfer/storage flask,
500 mL, joint: ST/NS 24/40, valve size 0-4 mm
Expand
View Pricing
SYNF484250
Synthware
™
solvent transfer/storage flask,
250 mL, joint: ST/NS 24/40, valve size 0-4 mm
Expand
View Pricing
SYNF484008
Synthware
™
solvent transfer/storage flask,
1000 mL, joint: ST/NS 24/40, valve size 0-8 mm
Expand
View Pricing
SYNF484508
Synthware
™
solvent transfer/storage flask,
500 mL, joint: ST/NS 24/40, valve size 0-8 mm
Expand
View Pricing
SYNF484000
Synthware
™
solvent transfer/storage flask,
1000 mL, joint: ST/NS 24/40, valve size 0-4 mm
Expand
View Pricing
SYNF484258
Synthware
™
solvent transfer/storage flask,
250 mL, joint: ST/NS 24/40, valve size 0-8 mm
Expand
View Pricing