Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Synthware
™
Schlenk tube with high vaccum valve
Products
Product Category
tubes (4)
Manufacturer
Capacity (mL)
OD (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
SYNF580810
Synthware
™
Schlenk tube with high vaccum valve,
capacity 10 mL, valve L 0-8 mm, O.D. × L 19 mm × 65 mm
Expand
View Pricing
SYNF580010
Synthware
™
Schlenk tube with high vaccum valve,
capacity 10 mL, valve L 0-4 mm, O.D. × L 22 mm × 55 mm
Expand
View Pricing
SYNF580020
Synthware
™
Schlenk tube with high vaccum valve,
capacity 20 mL, valve L 0-4 mm, O.D. × L 22 mm × 110 mm
Expand
View Pricing
SYNF580820
Synthware
™
Schlenk tube with high vaccum valve,
capacity 20 mL, valve L 0-8 mm, O.D. × L 19 mm × 30 mm
Expand
View Pricing