Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Synthware
™
fritted gas dispersion tube with 90 degree angle
Products
Product Category
tubes (4)
Manufacturer
Diameter (mm)
OD (mm)
Color
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
SYNF283045C
Synthware
™
fritted gas dispersion tube with 90 degree angle,
disc diam. 30 mm, porosity: coarse, stem O.D. × L 8 mm × 150 mm
Expand
View Pricing
SYNF282035M
Synthware
™
fritted gas dispersion tube with 90 degree angle,
disc diam. 20 mm, porosity: medium, stem O.D. × L 8 mm × 150 mm
Expand
View Pricing
SYNF284060X
Synthware
™
fritted gas dispersion tube with 90 degree angle,
disc diam. 40 mm, porosity: extra coarse, stem O.D. × L 8 mm × 150 mm
Expand
View Pricing
SYNF284060M
Synthware
™
fritted gas dispersion tube with 90 degree angle,
disc diam. 40 mm, porosity: medium, stem O.D. × L 8 mm × 150 mm
Expand
View Pricing