Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sure/Seal
™
bottles
Products
Product Category
bottles (4)
Brand
Sigma-Aldrich (4)
Manufacturer
Capacity (mL)
Cap
Color
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
Z406295
Sure/Seal
™
bottles,
capacity 125 mL, amber plastic-coated glass
Expand
View Pricing
Z406309
Sure/Seal
™
bottles,
capacity 1,000 mL, amber plastic-coated glass
Expand
View Pricing
Z567140
Sure/Seal
™
bottles,
capacity 500 mL, amber glass
Expand
View Pricing
Z406287
Sure/Seal
™
bottles,
capacity 1,000 mL, pkg of 20 ea
Expand
View Pricing