Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Super strength biohazard disposal bags
Products
Product Category
bags (5)
Manufacturer
Capacity (L)
Color
Package Size
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
BAF131651923
Super strength biohazard disposal bags,
width × H × thickness 19 in. (480 mm) × 23 in. (580 mm) × 0.05 mm (2 mil), pack of 200 ea, red
Expand
View Pricing
BAF131651419
Super strength biohazard disposal bags,
width × H × thickness 14 in. (360 mm) × 19 in. (480 mm) × 0.05 mm (2 mil), pack of 200 ea, red
Expand
View Pricing
BAF131653748
Super strength biohazard disposal bags,
width × H × thickness 37 in. (940 mm) × 48 in. (1220 mm) × 0.05 mm (2 mil), pack of 100 ea, red
Expand
View Pricing
BAF131652535
Super strength biohazard disposal bags,
width × H × thickness 25 in. (640 mm) × 35 in. (890 mm) × 0.05 mm (2 mil), pack of 200 ea
Expand
View Pricing
BAF131653138
Super strength biohazard disposal bags,
width × H × thickness 31 in. (790 mm) × 38 in. (960 mm) × 0.05 mm (2 mil), pack of 200 ea, red
Expand
View Pricing