Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Superdex
®
Increase 75
Products
Product Category
columns (3)
Brand
Sigma-Aldrich (3)
Manufacturer
pH Value
Product Line
Separation Technique
ID (mm)
Length (cm)
Volume (ml)
Column Material
Particle Size (µm)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
GE29-1487-21
Superdex
®
Increase 75,
10/300 GL, Cytiva 29-1487-21, column L × I.D. 30 cm × 10 mm, 9 μm avg. part. size
Expand
View Pricing
GE29-1487-23
Superdex
®
Increase 75,
3.2/300 GL, Cytiva 29-1487-23, column L × I.D. 30 cm × 3.2 mm, 9 μm avg. part. size
Expand
View Pricing
GE29-1487-22
Superdex
®
Increase 75,
5/150 GL, Cytiva 29-1487-22, column L × I.D. 15 cm × 5 mm, 9 μm avg. part. size
Expand
View Pricing