Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sudan IV
Synonym(s):
Biebrich scarlet R fat soluble, Fat Ponceau R or 4, Lipid Crimson, Oil Red IV, Scarlet Red Scharlach, Solvent Red 24, Sudan R or BB
Empirical Formula (Hill Notation):
C
24
H
20
N
4
O
CAS Number:
85-83-6
Molecular Weight:
380.44
EC Number:
201-635-8
Products
Product Category
stains (2)
Brand
Sigma-Aldrich (2)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Chemical Composition
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
198102
Sudan IV,
Certified Biological Stain Commission,Dye content ≥80 %
Expand
View Pricing
S4261
Sudan IV,
Dye content, ≥80%, certified by the Biological Stain Commission, powder
Expand
View Pricing
67386
Sudan IV,
analytical standard
Expand
View Pricing