Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Suba-Seal
®
silicone rubber septa
Products
Product Category
septa (5)
Brand
Sigma-Aldrich (5)
Color
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
Z279730
Suba-Seal
®
silicone rubber septa,
fits neck I.D., 8 mm, joint: ST/NS 10/30
Expand
View Pricing
Z279749
Suba-Seal
®
silicone rubber septa,
fits neck I.D., 14.0 mm, joint: ST/NS 14/20
Expand
View Pricing
Z279773
Suba-Seal
®
silicone rubber septa,
fits neck I.D., 22.0 mm, joint: ST/NS 24/40
Expand
View Pricing
Z279765
Suba-Seal
®
silicone rubber septa,
fits neck I.D., 19.0 mm, joint: ST/NS 19/22
Expand
View Pricing
Z279781
Suba-Seal
®
silicone rubber septa,
fits neck I.D., 27.0 mm, joint: ST/NS 29/42
Expand
View Pricing