Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Stirring bar retriever
Synonym(s):
Stirring bar retriever
Products
Product Category
stir bars (2)
Manufacturer
Diameter (mm)
Length (in)
Length (mm)
Color
Package Size
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
Z105090
Stirring bar retriever,
Rigid, PE, L 18 in.
Expand
View Pricing
Z105694
Stirring bar retriever,
Rigid, PE, L 12 in.
Expand
View Pricing
Z105708
Stirring bar retriever,
Flexible, PTFE, L × diam. 53 mm × 12.5 mm
Expand
View Pricing
BAH377740000
Stirring bar retriever,
white PTFE (flexible), diam. × L 12.5 in. (53 mm) × 2.0625 in. , magnet
Expand
View Pricing
BAF377810000
Stirring bar retriever,
Overall L 26 in. (65 cm)
Expand
View Pricing