Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Stearyl stearate
Synonym(s):
Stearic acid stearyl ester
Linear Formula:
CH
3
(CH
2
)
16
COO(CH
2
)
17
CH
3
CAS Number:
2778-96-3
Molecular Weight:
536.96
EC Number:
220-476-5
Products
Product Category
lipids (2)
fatty acids (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Biological Source
Formula Weight
Physical Form
Purity
Format
Quality Segment
Functional Group
Reference Material Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
85775
Stearyl stearate,
≥98.0% (GC)
Expand
View Pricing
46408
Stearyl stearate,
analytical standard
Expand
View Pricing