Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Stainless steel - AISI 316l
Products
Manufacturer
Physical Form
Length (m)
Length (mm)
OD (mm)
Filter Products
Showing 1-20 of 33
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF30719471
Stainless steel - AISI 316l,
powder, Fe/Cr18%/Ni 10%/Mo 3%, mean particle size 3 micron, weight 20 g, condition atomized
Expand
View Pricing
GF99421162
Stainless steel - AISI 316l,
tube, outside diameter 4.0 mm, Fe/Cr18%/Ni10%/Mo 3%, length 500 mm, wall thickness 0.55 mm
Expand
View Pricing
GF47468650
Stainless steel - AISI 316l,
tube, Fe/Cr18%/Ni 10%/Mo 3%, outside diameter 4.0 mm, length 100 mm, wall thickness 0.55 mm
Expand
View Pricing
GF38924310
Stainless steel - AISI 316l,
powder, Fe/Cr18%/Ni 10%/Mo 3%, max. particle size 45 micron, weight 50 g, condition atomized
Expand
View Pricing
GF42403205
Stainless steel - AISI 316l,
tube, Fe/Cr18%/Ni 10%/Mo 3%, outside diameter 2.1 mm, length 100 mm, wall thickness 0.25 mm
Expand
View Pricing
GF83597204
Stainless steel - AISI 316l,
powder, Fe/Cr18%/Ni10%/Mo 3%, 45 max. part. size (micron), weight 200 g, condition atomized
Expand
View Pricing
GF10480306
Stainless steel - AISI 316l,
tube, Fe/Cr18%/Ni 10%/Mo 3%, outside diameter 2.1 mm, length 500 mm, wall thickness 0.25 mm
Expand
View Pricing
GF67242843
Stainless steel - AISI 316l,
tube, outside diameter 1.65 mm, Fe/Cr18%/Ni10%/Mo 3%, length 1000 mm, wall thickness 0.13 mm
Expand
View Pricing
GF12594821
Stainless steel - AISI 316l,
wire, Fe/Cr18%/Ni 10%/Mo 3%, 0.5 mm diameter, size 1000 mm, condition straight wire
Expand
View Pricing
GF16783768
Stainless steel - AISI 316l,
foil, Fe/Cr18%/Ni 10%/Mo 3%, thickness 1.2 mm, size 100 × 100 mm, temper annealed
Expand
View Pricing
GF58725995
Stainless steel - AISI 316l,
tube, Fe/Cr18%/Ni 10%/Mo 3%, outside diameter 4.0 mm, length 200 mm, wall thickness 0.55 mm
Expand
View Pricing
GF59218881
Stainless steel - AISI 316l,
tube, Fe/Cr18%/Ni 10%/Mo 3%, outside diameter 2.1 mm, length 1000 mm, wall thickness 0.25 mm
Expand
View Pricing
GF63977439
Stainless steel - AISI 316l,
tube, outside diameter 1.65 mm, Fe/Cr18%/Ni10%/Mo 3%, length 500 mm, wall thickness 0.13 mm
Expand
View Pricing
GF83004544
Stainless steel - AISI 316l,
foil, thickness 0.25 mm, Fe/Cr18%/Ni10%/Mo 3%, length 5 m, coil width 12 mm
Expand
View Pricing
GF67614228
Stainless steel - AISI 316l,
powder, 45 max. part. size (micron), Fe/Cr18%/Ni10%/Mo 3%, weight 100 g, condition atomized
Expand
View Pricing
GF84216641
Stainless steel - AISI 316l,
rod, 5.0 mm diameter, Fe/Cr18%/Ni10%/Mo 3%, length 1000 mm
Expand
View Pricing
GF38479821
Stainless steel - AISI 316l,
tube, Fe/Cr18%/Ni 10%/Mo 3%, outside diameter 4.0 mm, length 1000 mm, wall thickness 0.55 mm
Expand
View Pricing
GF94811092
Stainless steel - AISI 316l,
tube, Fe/Cr18%/Ni10%/Mo 3%, length 200 mm, outside diameter 2.1 mm, wall thickness 0.25 mm
Expand
View Pricing
GF73205287
Stainless steel - AISI 316l,
powder, 45 max. part. size (micron), Fe/Cr18%/Ni10%/Mo 3%, weight 500 g, condition atomized
Expand
View Pricing
GF86268606
Stainless steel - AISI 316l,
non-woven fabric, Fe/Cr18%/Ni10%/Mo 3%, weight.m-2 -50g, size 150 × 150 mm, thickness 0.060 mm
Expand
View Pricing
Page 1 of 2
Page 1 of 2