Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
SS-174 PTFE-Faced Septa
Products
Product Category
septa (5)
Brand
Supelco (3)
Diameter (mm)
Color
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
22731
SS-174 PTFE-Faced Septa,
diam. 11 mm ( 7/16 in.)
Expand
View Pricing
22656
SS-174 PTFE-Faced Septa,
diam. 9.5 mm ( 3/8 in.), pkg of 100 ea
Expand
View Pricing
22654
SS-174 PTFE-Faced Septa,
diam. 6 mm ( 1/4 in.)
Expand
View Pricing
22657
SS-174 PTFE-Faced Septa,
diam. 12.5 mm ( 1/2 in.)
Expand
View Pricing
22647
SS-174 PTFE-Faced Septa,
diam. 10 mm ( 13/32 in.)
Expand
View Pricing