Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Soya DAS-44406-6 GMO standard ERM-BF436
Synonym(s):
DAS-44406-6 soya seed
Products
Manufacturer
Agency
Format
Application
Reference Material Type
Special Grade
certified reference material (5)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
ERMBF436C
Soya DAS-44406-6 GMO standard ERM-BF436,
ERM
®
, certified reference material, nominal 0.1% GMO
Expand
View Pricing
ERMBF436E
Soya DAS-44406-6 GMO standard ERM-BF436,
ERM
®
, certified reference material, nominal 10% GMO
Expand
View Pricing
ERMBF436A
Soya DAS-44406-6 GMO standard ERM-BF436,
ERM
®
, certified reference material, blank
Expand
View Pricing
ERMBF436B
Soya DAS-44406-6 GMO standard ERM-BF436,
ERM
®
, certified reference material, nominal 100% GMO
Expand
View Pricing
ERMBF436D
Soya DAS-44406-6 GMO standard ERM-BF436,
ERM
®
, certified reference material, nominal 1% GMO
Expand
View Pricing