Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sorenson
™
low binding Ultra G microtips
Products
Product Category
pipette tips (4)
Manufacturer
Color
Package Size
Sterility
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
Z719498
Sorenson
™
low binding Ultra G microtips,
maximum volume 10 μL, pack of 1000 ea (Bag), non-sterile
Expand
View Pricing
Z719501
Sorenson
™
low binding Ultra G microtips,
maximum volume 10 μL, pack of 96 ea Cell Rack, non-sterile
Expand
View Pricing
Z719528
Sorenson
™
low binding Ultra G microtips,
maximum volume 10 μL, pack of 96 ea CellRack, sterile
Expand
View Pricing
Z719536
Sorenson
™
low binding Ultra G microtips,
maximum volume 10 μL, pkg of 960 tips QuickRack, non-sterile
Expand
View Pricing