Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium thiosulfate solution
Synonym(s):
Sodium thiosulfate solution
Linear Formula:
Na
2
S
2
O
3
CAS Number:
7772-98-7
Molecular Weight:
158.11
Products
Product Category
detergents (4)
histology special stains (1)
Brand
Sigma-Aldrich (5)
Formula Weight
Physical Form
Quality Segment
Application
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
28-4540
Sodium thiosulfate solution,
0.1 M
Expand
View Pricing
28-4560
Sodium thiosulfate solution,
0.01 M
Expand
View Pricing
HT1005
Sodium thiosulfate solution,
2 g/dL in deionized water
Expand
View Pricing
28-4550
Sodium thiosulfate solution,
0.025 M
Expand
View Pricing
28-4530
Sodium thiosulfate solution,
1 M
Expand
View Pricing