Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium metasilicate nonahydrate
Empirical Formula (Hill Notation):
Na
2
O
3
Si · 9H
2
O
CAS Number:
13517-24-3
Molecular Weight:
284.20
EC Number:
229-912-9
Products
Product Category
salts (3)
Brand
Sigma-Aldrich (3)
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
S4392
Sodium metasilicate nonahydrate,
≥98%
Expand
View Pricing
S5904
Sodium metasilicate nonahydrate,
BioReagent, suitable for plant cell culture, ≥98%
Expand
View Pricing
28-3360
Sodium metasilicate nonahydrate,
CP
Expand
View Pricing