Skip to Content
Merck
Sodium dodecyl sulfate solution BioUltra, 10% in H2O

Sodium dodecyl sulfate solution

Sodium dodecyl sulfate solution BioUltra, 10% in H2O
Synonym(s):
Lauryl sulfate sodium salt, SDS, Sodium lauryl sulfate solution, Sodium n-Dodecyl Sulfate, 20% Solution (w/v)
Linear Formula:
CH3(CH2)11OSO3Na
CAS Number:
Molecular Weight:
288.38

Products

Product Category
Brand
Sustainability
Color
Feature
Formula Weight
pH Value
Physical Form
Product Line
Quality Segment
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing