Skip to Content
Merck
Sodium creatine phosphate dibasic tetrahydrate ≥98.0% (NT)

Sodium creatine phosphate dibasic tetrahydrate

Sodium creatine phosphate dibasic tetrahydrate ≥98.0% (NT)
Synonym(s):
Creatine phosphate disodium salt tetrahydrate
Linear Formula:
Na2O3PNHC(=NH)N(CH3)CH2COOH · 4H2O
CAS Number:
Molecular Weight:
327.14
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing