Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium bis(trifluoromethanesulfonyl)imide solution
Products
Product Category
electrolytes (2)
Brand
Sigma-Aldrich (2)
Sustainability
Feature
Physical Form
Quality Segment
Application
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
936014
Sodium bis(trifluoromethanesulfonyl)imide solution,
in ethylene carbonate, dimethylcarbonate, and ethylmethylcarbonate, 1.0 M NaTFSI in EC/DMC/EMC=1:1:1 (v/v/v), battery grade
Expand
View Pricing
936006
Sodium bis(trifluoromethanesulfonyl)imide solution,
in ethylene carbonate, dimethyl carbonate, 1.0 M NaTFSI in EC/DMC=50/50 (v/v), battery grade
Expand
View Pricing