Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Smart SPME Fiber Assembly
Products
Brand
Supelco (12)
Sustainability
Feature
Length (cm)
Color
d
f
(µm)
Size
Gauge
Technique
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
548653-U
Smart SPME Fiber Assembly,
DVB/CAR/PDMS, for use with PAL 3, Series II Smart autosampler, Nitinol-core (NIT), 23 ga, 50 μm (DVB layer), 30 μm (CAR/PDMS layer)
Expand
View Pricing
548551-U
Smart SPME Fiber Assembly,
CAR/PDMS, for use with PAL 3, Series II Smart autosampler, Nitinol-core (NIT), 23 ga, 85 μm (CAR/PDMS)
Expand
View Pricing
548575-U
Smart SPME Fiber Assembly,
PDMS, for use with PAL 3, Series II Smart autosampler, Nitinol-core (NIT), 23 ga, 100 μm (PDMS)
Expand
View Pricing
548550-U
Smart SPME Fiber Assembly,
CAR/PDMS, for use with PAL 3, Series 2 Smart autosampler, Nitinol-core (NIT), 23 ga, 75 μm (CAR/PDMS)
Expand
View Pricing
548675-U
Smart SPME Fiber Assembly,
DVB/CAR/PDMS, for use with PAL 3, Series II Smart autosampler, Nitinol-core (NIT), 23 ga, 50 μm (DVB layer), 30 μm (CAR/PDMS layer)
Expand
View Pricing
548652-U
Smart SPME Fiber Assembly,
PA, for use with PAL 3, Series II Smart autosampler, Nitinol-core (NIT), 23 ga, 85 μm (PA)
Expand
View Pricing
548576-U
Smart SPME Fiber Assembly,
PDMS/DVB, for use with PAL 3, Series II Smart autosampler, Nitinol-core (NIT), 23 ga, 65 μm(PDMS/DVB)
Expand
View Pricing
548650-U
Smart SPME Fiber Assembly,
PDMS/DVB, for use with PAL 3, Series II Smart autosampler, Nitinol-core (NIT), 23 ga, 65 μm (PDMS/DVB)
Expand
View Pricing
548651-U
Smart SPME Fiber Assembly,
PDMS/DVB, Overcoated, for use with PAL 3, Series II Smart autosampler, Nitinol-core (NIT), 23 ga, 75 μm(Coating thickness includes 65 μm coating + 10 μm OC (overcoating)
Expand
View Pricing
548552-U
Smart SPME Fiber Assembly,
PDMS, for use with PAL 3, Series II Smart autosampler, Nitinol-core (NIT), 23 ga, 7 μm (PDMS)
Expand
View Pricing
548676-U
Smart SPME Fiber Assembly,
PEG, for use with PAL 3, Series 2 Smart autosampler, Nitinol-core (NIT), 23 ga, 60 μm (PEG)
Expand
View Pricing
548553-U
Smart SPME Fiber Assembly,
PDMS, for use with PAL 3, Series II Smart autosampler, Nitinol-core (NIT), 23 ga, 30 μm (PDMS)
Expand
View Pricing