Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Single-bulb ball-tube
Products
Product Category
tubes (18)
Brand
Sigma-Aldrich (18)
Capacity (mL)
Filter Products
Showing 1-18 of 18
Compare
Product Number
Description
Pricing
Z101915
Single-bulb ball-tube,
capacity 250 mL, joint: ST/NS 24/40, plain
Expand
View Pricing
Z101931
Single-bulb ball-tube,
capacity 1,000 mL, joint: ST/NS 24/40, plain
Expand
View Pricing
Z101923
Single-bulb ball-tube,
capacity 500 mL, joint: ST/NS 24/40, plain
Expand
View Pricing
Z174300
Single-bulb ball-tube,
capacity 2,000 mL, joint: ST/NS 24/40, plain
Expand
View Pricing
Z100587
Single-bulb ball-tube,
capacity 50 mL, joint: ST/NS 14/20, plain
Expand
View Pricing
Z100617
Single-bulb ball-tube,
capacity 500 mL, joint: ST/NS 14/20, plain
Expand
View Pricing
Z219746
Single-bulb ball-tube,
capacity 10 mL, joint: ST/NS 14/20, w/drip-tip
Expand
View Pricing
Z100579
Single-bulb ball-tube,
capacity 25 mL, joint: ST/NS 14/20, plain
Expand
View Pricing
Z100595
Single-bulb ball-tube,
capacity 100 mL, joint: ST/NS 14/20, plain
Expand
View Pricing
Z219770
Single-bulb ball-tube,
capacity 100 mL, joint: ST/NS 14/20, w/drip-tip
Expand
View Pricing
Z219762
Single-bulb ball-tube,
capacity 50 mL, joint: ST/NS 14/20, w/drip-tip
Expand
View Pricing
Z219800
Single-bulb ball-tube,
capacity 250 mL, joint: ST/NS 24/40, w/drip-tip
Expand
View Pricing
Z219819
Single-bulb ball-tube,
capacity 500 mL, joint: ST/NS 24/40, w/drip-tip
Expand
View Pricing
Z219797
Single-bulb ball-tube,
capacity 500 mL, joint: ST/NS 14/20, w/drip-tip
Expand
View Pricing
Z219827
Single-bulb ball-tube,
capacity 1,000 mL, joint: ST/NS 24/40, w/drip-tip
Expand
View Pricing
Z219789
Single-bulb ball-tube,
capacity 250 mL, joint: ST/NS 14/20, w/drip-tip
Expand
View Pricing
Z100609
Single-bulb ball-tube,
capacity 250 mL, joint: ST/NS 14/20, plain
Expand
View Pricing
Z219754
Single-bulb ball-tube,
capacity 25 mL, joint: ST/NS 14/20, w/drip-tip
Expand
View Pricing