Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Silver diethyldithiocarbamate
Synonym(s):
Silver diethyldithiocarbamate, DETC, Diethyldithiocarbamic acid silver salt
Linear Formula:
(C
2
H
5
)
2
NCSSAg
CAS Number:
1470-61-7
Molecular Weight:
256.14
EC Number:
216-003-7
Products
Product Category
catalysts (1)
inhibitors (1)
ligands (1)
Brand
Sigma-Aldrich (3)
Supelco (1)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reagent Type
Core
Special Grade
ACS reagent (2)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
1.01515
Silver diethyldithiocarbamate,
for analysis EMSURE
®
,Reag. Ph Eur
Expand
View Pricing
D3132
Silver diethyldithiocarbamate,
Expand
View Pricing
D93503
Silver diethyldithiocarbamate,
ACS reagent, 99%
Expand
View Pricing
85180
Silver diethyldithiocarbamate,
ACS reagent, for the determination of As, ≥99.0%
Expand
View Pricing