
Sildenafil citrate

Synonym(s):
Sildenafil citrate salt, 1-[[3-(4,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]-4-methylpiperazine citrate salt, 5-[2-Ethoxy-5-(4-methylpiperazin-1-yl)sulfonylphenyl]-1-methyl-3-propyl-4H-pyrazolo[5,4-e]pyrimidin-7-one citrate salt, Revatio, UK-92,480
Empirical Formula (Hill Notation):
C22H30N6O4S ·C6H8O7
CAS Number:
Molecular Weight:
666.70
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing