Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sigma-Aldrich
®
TLC Plates
Products
Product Category
plates (4)
Brand
Supelco (4)
Manufacturer
Length (cm)
Quality Segment
Fluorescence
Matrix Material
Support Material
Particle Size (µm)
Pore Size (Å)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
Z122785
Sigma-Aldrich
®
TLC Plates,
silica gel 60 matrix
Expand
View Pricing
Z193291
Sigma-Aldrich
®
TLC Plates,
silica gel matrix, L × W 20 cm × 20 cm
Expand
View Pricing
Z122726
Sigma-Aldrich
®
TLC Plates,
silica gel 60 matrix, L × W 20 cm × 20 cm, fluorescent indicator
Expand
View Pricing
Z122882
Sigma-Aldrich
®
TLC Plates,
PEI-cellulose, matrix
Expand
View Pricing